SP Bel-Art H-B® DURAC® Four-Channel Electronic Timer with White Board and Certificate of Calibration, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
B61700-3700
10061-386CS 601.83 USD
10061-386
SP Bel-Art H-B® DURAC® Four-Channel Electronic Timer with White Board and Certificate of Calibration, Bel-Art Products, a part of SP
Timers Digital Timers
This four-channel timer has a built-in white board and dry erase marker with eraser tip that helps keep track of time and other critical information.
  • Audible alarms
  • Magnetic backing to attach to metal surfaces
  • Multiple timing functions
Each channel a switchable timing capacity of 23 hours: 59 minutes or 59 minutes, and last time set memory. It programs in hours/minutes and displays in hours/minutes/seconds. Up or down counting is possible, and elapsed time after countdown is displayed. A key press confirmation feature gives an audible indication ofMore Product Information