Rt®-QS-BOND Columns (fused silica PLOT), Restek
Supplier: Restek
19737 19740 19738 19767 19768 19739
10058-538EA 679.02 USD
10058-538 10058-540 10803-206 10848-970 10848-972 10851-502
Rt®-QS-BOND Columns (fused silica PLOT), Restek
Chromatography Columns
Porous polymers are unique, highly retentive stationary phases with a wide application range that are able to elute both polar and nonpolar compounds.
  • Intermediate polarity porous polymer PLOT column incorporating low 4-vinylpyridine
  • Separates ethane, ethylene, and acetylene to baseline
  • Stable to 250 °C
  • Porous divinylbenzene homopolymer
These columns are very hydrophobic, so water has no impact on retention times and even elutes as a good chromatographic peak. The Q-BOND is Restek's most nonpolar and widely used porous polymer column; functionalMore Product Information