VWR® Disposable Petri Dishes
Supplier: VWR International
3308 3561 3515 3505 3550 3306 3310 3305 3501 3502 3503 3504 3562 3572 3573 3511 3525 3508 3516 3517 3518 3519 3501A 3571WI 3561WI 3520 3572WI 3571 3574 3577 3509 3552 3596 3506 3565 3548NI 3527 3535 3521
25384-062CS 268.12 USD
25384-062 25384-088 25384-090 25384-092 25384-093 25384-164 25384-166 25384-168 25384-302 25384-304 25384-306 25384-308 25384-310 25384-312 25384-314 25384-324 25384-326 25384-332 25384-342 25384-344 25384-346 25384-348 89022-320 89038-968 89038-970 89107-632 89137-760 25384-094 25384-316 25384-338 25384-318 25384-320 25384-322 25384-328 89230-472 10053-704 10799-140 10799-192 10799-194
VWR® Disposable Petri Dishes
Culture Dishes Petri Dishes
Clearly superior, high-quality Petri dishes are manufactured from virgin crystal grade polystyrene for optimum flatness and high optical clarity.
  • Virgin crystal grade polystyrene construction
  • Available in varied sizes and styles for specialized and routine applications
  • Stacking ring and slip-ring features available
  • Supplied sterile
VWR® Petri dishes are available in a variety of sizes and styles to accommodate both specialized and routine needs. The stacking ring feature, on some models, facilitates positive stacking while the slip-ring feature, onMore Product Information