Anti-JNK1 Mouse Monoclonal Antibody [clone: 5D10]

Supplier: Enzo Life Sciences
905-649-100
10051-914EA 610.4 USD
10051-914
Anti-JNK1 Mouse Monoclonal Antibody [clone: 5D10]
Antibodies
The Stress-activated protein kinases alpha and gamma (SAPKalpha and gamma), also know as c-Jun NH2-terminal kinases 1 and 2 (Jnk1/2) are serine/ threonine protein kinases belonging to the MAP kinase family. The extracellular stimuli leading to cell stress include inflammatory cytokines (via Rho and Rac), growth factors, protein synthesis inhibitors and osmotic stress amongst others. Both Jnk1 and Jnk2 must be dually phosphorylated in the activation loop, at threonine 183 and tyrosine 185, to achieve full enzymatic activity.

Type: Primary
Antigen: JNK1
Clonality: Monoclonal
Clone: 5D10
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR