Mouse IgG2a Monoclonal Isotype Control [clone: C1.18.4] (PE (Phycoerythrin))

Supplier: Tonbo Biosciences
50-4724-U100
10051-332EA 106.7 USD
10051-332
Mouse IgG2a Monoclonal Isotype Control [clone: C1.18.4] (PE (Phycoerythrin))
Antibodies
The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.

Type: Control
Antigen: C1.18.4
Clonality: Monoclonal
Clone: C1.18.4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR