Sodium carbonate, anhydrous ACS

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Calcined soda, Carbonic acid disodium salt

21135
100504-276EA 89.22 USD
100504-276
Sodium carbonate, anhydrous ACS
Sodium carbonate

Na2CO3 F.W. 105.99 CAS 497-19-8

Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600 °C (2912 °F)
Melting Pt: 851 °C (1564 °F)
Density: 2.532 g/cm³ (21.13 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71793d43eac72db07a92291d463220982ec0fec5&vid=71793d43eac72db07a92291d463220982ec0fec5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Ammonium hydroxide ppt 0.01%
Arsenic 1 ppm
Ca and Mg 0.01%
Chloride 0.001%
Heavy metals 5 ppm
Insoluble matters 0.01%
Iron 5 ppm
Loss on heating (280 °C) 1.0%
Nitrogen compounds 0.001%
Phosphate 0.001%
Potassium 0.005%
Silica (SiO₂) 0.005%
Sulfur compounds (as S) 0.003%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR