Polyethylene glycol 3,350 waxy

Supplier: Electron Microscopy Sciences

Synonyms: PEG 3,350, PEG 3350

19760
100504-216EA 162.62 USD
100504-216
Polyethylene glycol 3,350 waxy
Polyethylene glycol 3,350

An embedding medium for microscopy and histochemistry

Formula: H(OCH₂CH₂)nOH
MW: 3350 g/mol
Density: 1.0926 (60 °C)
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR