Benzoin methyl ether ≥98% UV catalyst

Supplier: Electron Microscopy Sciences
Danger

Synonyms: 1,2-Diphenyl-2-methoxyethan-1-one, 2-Methoxy-1,2-diphenylethanone

11290
100503-686EA 80.63 USD
100503-686
Benzoin methyl ether ≥98% UV catalyst
Benzoin methyl ether

[alpha-Ethoxy-alpha-phenylacetophenone) C6H5CH(OCH3)COC6H5 F.W. 226.28 CAS 3524-62-7 Assay.98% m.p. 47 - 49°C

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00008492
CAS Number: 3524-62-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=531c19c79a713588d563469574c16b85469b5226&vid=d5aa4f080e720bd38241a26bea3f1a4dd92928e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay >98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR