Rat IgG1 Monoclonal Isotype Control [clone: HRPN] (APC (Allophycocyanin))

Supplier: Tonbo Biosciences
20-4301-U100
10050-604EA 123.2 USD
10050-604
Rat IgG1 Monoclonal Isotype Control [clone: HRPN] (APC (Allophycocyanin))
Antibodies
The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.

Type: Control
Antigen: HRPN
Clonality: Monoclonal
Clone: HRPN
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR