Histo Orientator, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63205
100496-160EA 1098.91 USD
100496-160
Histo Orientator, Electron Microscopy Sciences
Heaters Hot Plates
The convenient 51×51 mm (2×2") heated surface can be used for either 'de-wrinkling' of paraffin sections or for the 'rapid attachment' of fresh frozen sections. A quick touch of the slide to the hot plate will remove wrinkles and bubbles on sections. Also keep a unit on your cryostat to ensure solid attachment of frozen sections to the slide which prevents loosening of the tissue during agitation in hand staining.
  • The unit measures: 4½×2¾×3¼" (114×70×82 mm)
  • Electrical: 120 V, 60 Hz, 12 W
  • Temperature range: ambient to 65 °C
Order Now
SPECIFICATIONS
Environmentally Preferable