Veco Parallel Bar with Divider, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
R300D-CU R100AS-CU R100D-NI R100AS-NI R200D-CU R200D-NI R300D-NI R150D-CU R150D-NI
100495-998EA 40.7 USD
100495-998 100495-964 100495-972 100495-966 100495-990 100495-992 100496-000 100495-982 100495-984
Veco Parallel Bar with Divider, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco Grids offer superior handling characteristics.
  • With 0.8 mil thickness
  • Measures 3.05 mm in diameter
Veco Grids are the most rigid grids available. Material: Copper (Cu); Nickel (Ni).
Order Now
SPECIFICATIONS
Environmentally Preferable