Negative Carriers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74868-40
100494-086EA 289.64 USD
100494-086 100494-080 100494-078 100494-098 100494-096 100494-072 100494-088 100494-082 100494-084
Negative Carriers, Electron Microscopy Sciences
Film Processing Supplies
Universal glass-type masking negative carriers features four individually adjustable masking blades and removable glass inserts
Glassless, sandwich type negative carriers are rofessional quality, with black, anodized aluminum construction. Hinged for quick handling.
Universal glass-type masking negative carriers features four individually adjustable masking blades and removable glass inserts. Accepts accessory anti-Newton glass. Top and bottom glass can be removed to convert it to a single-size glassless carrier by means of lock-in inserts.
Accepts accessory anti-Newton glass. Top andMore Product Information