Stop Watch: Large Display, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72091
100491-924EA 69.31 USD
100491-924
Stop Watch: Large Display, Electron Microscopy Sciences
Timers Digital Stopwatches
Very accurate with a click stop control button.
  • Large LCD display
  • Rugged ABS plastic case
It counts up to 60 minutes in ¹/₁₀₀th second resolution. It also has a split interval clock and calculator functions. Measures: 3×2½×¼".
Weight: 2.6 oz.
Información de suministro: Comes complete with a leatherette case.