EMS Domino Racks, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70620 70621
100491-638EA 57.87 USD
100491-638 100496-274
EMS Domino Racks, Electron Microscopy Sciences
Racks Film Racks
The EMS Domino rack is "U" shaped and made from an aircraft alloy sheet stock with serial perforations; thermally bonded spaceage copolymer.
  • Reduces the film and section contamination to negligible levels
  • Allows the sections within the slot to dry flat and wrinkle free
The formvar film cast on the rack will stretch across a series of smooth edged holes forming a flat, wrinkle free film that is ready for grid mounting.
Moran, D.T., and Rowley, J.C., (1987). Biological Specimen Preparation for Correlative Light and Electron Microscopy in Biology:More Product Information