Slide Duplicating With Cover Slip Mold, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70172
100491-610EA 34.01 USD
100491-610
Slide Duplicating With Cover Slip Mold, Electron Microscopy Sciences
Moulds Moulds for Histology
The same design as slide duplicating mold, but with the addition of three round cavities.
Cavities are 17 mm in diameter and 1 mm deep.