Lab-Tek® Chambered Coverglass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70377-22 70377-81 70377-42 70377-82 70378-81 70377-41 70378-82 70378-11 70377-11 70377-12 70378-12 70378-21 70378-42 70377-21 70378-41 70378-22
100489-100PK 2212.88 USD
100489-100 100489-104 100489-102 100489-106 100489-112 100490-938 100500-082 100501-814 100501-210 100489-096 100500-076 100489-108 100500-080 100489-098 100489-110 100500-078
Lab-Tek® Chambered Coverglass, Electron Microscopy Sciences
Cover Glasses
Lab-Tek® and Lab-Tek®II Chambered Coverglass products are intended for high magnification examination of living cells using an inverted microscope.
The growth surface of the Lab-Tek Chambered Coverglass products are #1 Borosilicate Coverglass from American origin. The Lab-Tek¨ II Chambered Coverglass products are #1.5 Borosilicate Coverglass of German origin. The thin glass growth surface allows optimal optical clarity when viewing living cells with high resolution of an inverted microscope. The media chambers are available in 1, 2, 4, and 8 well formats and cannot be removed from theMore Product Information