PTFE Printed Slides, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63414-10 63422-06 63424-06 63423-05 63415-12 63425-05 63429-04 63415-32 63421-11 63416-08 63419-14 63420-05 63416-15 63419-12 63430-04 63414-13 63419-VIR 63423-08 63415-08 63419-08 63420-08 63422-11 63434-02 63417-13 63421-10
100490-316PK 131.26 USD
100490-316 100488-900 100488-904 100490-328 100490-334 100488-876 100488-906 100488-898 100488-908 100490-320 100490-332 100488-880 100488-892 100488-894 100488-882 100488-888 100488-890 100488-910 100490-318 100490-326 100490-336 100488-874 100488-884 100488-886 100488-896 100488-902 100488-912 100490-322 100490-330
PTFE Printed Slides, Electron Microscopy Sciences
Slides Microscope Slides
These slides are extremely hydrophobic, acetone proof, chemical resistant and autoclavable. They are perfect for controlling cross-contamination in staining, immunolabeling and immunofluorescence tests.
  • Super hydrophobic mask
  • Clean, wettable wells
  • A bioadhesive surface treatment to enhance cell and tissue attachment
The well surface is 100% wettable, which enhances cell attachment and increases its spreading capabilities within the well. These slides minimize the use of expensive purified antigens and antibodies. The slides measure 3 x 1" (76 x 26 x 1More Product Information