SOLA Solid Phase Extraction Plates and Cartridges, Thermo Scientific
Supplier: Thermo Scientific
60309-001 60309-002 60309-003 60309-004 60309-005 60209-001 60209-002 60209-003 60209-004 60209-005 60509-001 60509-002 60509-003 60509-004 60509-005
10038-926EA 465.89 USD
10038-926 10038-928 10038-930 10038-932 10038-934 10119-788 10119-790 10119-792 10119-794 10119-796 76457-320 76457-322 76457-324 76457-326 76457-328
SOLA Solid Phase Extraction Plates and Cartridges, Thermo Scientific
Microplates Solid Phase Extraction Plates (SPE)
SOLA is a state of the art fritless polymeric SPE that reduces, channeling, voiding, packing inconsistencies and reduces the need for expense retesting of critical samples.
  • Fritless
  • Polymeric
  • Consistent bed
  • More reproducible
  • Greater selectivity
  • Cleaner
Prevent sample failures in bioanalytical workflows by using SOLA SPE.
SOLA and SOLAu SPE products help prevent costly reanalysis of bioanalytical samples by preventing blocking, voiding, and channeling during the SPE sample preparation process. The fritless polymeric technology eliminates the issuesMore Product Information