VWR® UV-3100PC, V-3000-PC Spectrophotometers, UV-Vis Scanning and Vis
Supplier: VWR International
10037-438 10037-440
10037-438CS 4900.27 USD
10037-438 10037-440
VWR® UV-3100PC, V-3000-PC Spectrophotometers, UV-Vis Scanning and Vis
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Two fully scanning spectrophotometers for advanced measurements and quality control.
  • Large LCD display (320×240 pixels)
  • Full scan mode
  • Easily accessible USB port for data import and export
  • Supplied application software 'UV-Vis Analyst' allows PC control of the spectrophotometer
  • Methods for PC control or standalone include: Basic mode, quantitative, wavelength scan, kinetics, multi-wavelength, DNA/protein
  • The range of accessories includes an automatic 4-cell changer and Peltier thermostatted single cell holder
Reliable, robust and easy to useMore Product Information