Filaments, Sutter Instruments
Supplier: Sutter Instruments Company
FB315B FB215B FT315B FB220B FB245B FB330B FB255B FB320B FT320B FB230B FT330B FT345B
100357-906EA 71.92 USD
100357-906 100357-896 100357-918 100357-898 100357-902 100357-910 100357-904 100357-908 100357-920 100357-900 100357-922 100357-924
Filaments, Sutter Instruments
Puller and Inserter Tools
Appropriate filament selection depends on your research application.
Box Filaments are recommended for small to large diameter borosilicate glass, double barreled, or aluminosilicate glass. Box filaments are particularly suitable for patch pipettes, slice preparations, and for sharp electrodes and microinjection pipettes where gradual parallel walls would aid cell penetration. When using a box filament, the size of the square box should be approximately 1.0 to 1.5 mm larger than the outside diameter of the glass that you will be using.
Trough Filaments are excellent general purposeMore Product Information