PeridoxRTU® Sporicidal Disinfectant and Cleaner, Contec
Supplier: CONTEC, INC. MX
CR85336 CR85335 CR85335IR CR85336IR CRPERTRIGEB
10032-832CS 413.43 USD
10032-832 10032-834 10148-324 10746-534 76460-620
PeridoxRTU® Sporicidal Disinfectant and Cleaner, Contec
Cleaning Agents Disinfectants
PeridoxRTU® is a broad spectrum EPA registered sporicide, bactericide, virucide, tuberculocide, and fungicide one-step disinfectant and hard surface cleaner with a 3-minute Sporicide claim at 99.9999% efficacy. It contains no alcohol or bleach and, unlike most other disinfectants, leaves minimal residue on surfaces.
  • EPA registered sporicidal disinfectant and cleaner
  • 3-minute sporicidal claim at 99.9999% efficacy
  • Does not contain bleach, alcohol, phenols, or quaternary ammonium compounds
  • Excellent material compatibility will not damage most surfaces in
More Product Information