Incubators, Refrigerated with Peltier Technology, KT Series, BINDER
Supplier: Binder
9020-0312 9020-0310 9020-0314
10031-394EA 6586.8 USD
10031-394 10823-814 75838-570
Incubators, Refrigerated with Peltier Technology, KT Series, BINDER
Incubators Cooled Incubators
The KT refrigerated incubators with Peltier technology combine outstanding performance with environmental friendliness and provide sample protection.
These units feature the electronically controlled APT.line™ preheating chamber provide even horizontal airflow on both sides for homogenous temperature distribution along with long term stability. The units utilize Peltier refrigeration technology for finely controlled cooling from 4° to 100°C as well as a powerful conventional tubular heating system.
The disinfection routine is performed at 100°C in a high quality stainless steel innerMore Product Information