Radleys Cooled Carousel 6 Plus™ Reaction Station, Heidolph®
Supplier: Heidolph NA, LLC
015850490 015850510
10031-072EA 15523.6 USD
10031-072 10031-074
Radleys Cooled Carousel 6 Plus™ Reaction Station, Heidolph®
Stirrers Magnetic Hotplate Stirrers
The Cooled Carousel 6 Plus™ Reaction Station allows chemists to perform sub-ambient reactions in a range of flasks from 5mL to 250mL with the option of an inert, moisture-free atmosphere.
The station allows simultaneous stirring and cooling reactions from ambient down to –70°C (–94°F) using up to six round-bottom flasks in 5mL, 10mL, 25mL, 50mL, 100mL, 170mL, and 250mL sizes. As with all Radleys Carousel Stations, any combination of these flask sizes can be used in the Cooled Carousel, which fits on a standard hotplate stirrer.
Cooled Carousel design and technology provides evenMore Product Information