n-Dodecyl-β-D-maltoside ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside

0411281GM
100294-488EA 141.23 USD
100294-488
n-Dodecyl-β-D-maltoside ≥99%
n-Dodecyl-β-D-maltoside
Formula: C₂₄H₄₆O₁₁
MW: 510.6 g/mol
MDL Number: MFCD00043012
CAS Number: 69227-93-6
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7888724c33362326b178a9a214cb3b49cd3c16fc&vid=7888724c33362326b178a9a214cb3b49cd3c16fc&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bf74a55d8e1f69041856e43234c6b7538c77fd1&vid=0bf74a55d8e1f69041856e43234c6b7538c77fd1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR