VWR® Carboy Dolly
Supplier: VWR International
10029-542
10029-542EA 1038.05 USD
10029-542
VWR® Carboy Dolly
Carts
The VWR® Carboy Dolly features stainless steel construction, allowing it to carry carboys or contents up to 90kg (200lbs.).
  • Stainless steel construction with polyurethane wheels
  • Versatile and maneuverable
Four white polyurethane wheels pivot for maximum versatility and maneuverability, without damaging floors. The Carboy Dolly is specifically designed for use with 40L and 75L VWR® Carboys.