Forced Air Circulation Oven Units, Memmert
Supplier: Memmert
UF30, 115 V UF30, 230 V UF30PLUS,115 UF30PLUS,230 UF55, 115 V UF55, 230 V UF55PLUS230 UF75, 115 V UF75, 230 V UF75PLUS115 UF75PLUS230 UF110, 115 V UF110, 230 V UF110PLUS115 UF110PLUS230 UF160, 115 V UF160, 230 V UF160PLUS115 UF160PLUS230 UF260, 115 V UF260, 230 V UF260PLUS115 UF260PLUS230 UF450230,3PH UF450PLUS3PH UF750230,3PH UF750PLUS3PH UF1060230V3PH UF1060PLUS3PH
10029-024EA 3146 USD
10029-024 10029-026 10029-028 10029-030 10029-032 10029-034 10029-036 10029-038 10029-056 10029-058 10029-060 10029-062 10029-064 10029-066 10029-068 10029-070 10029-072 10029-074 10029-076 10029-078 10029-080 10029-082 10029-084 10029-086 10029-088 10029-090 10029-092 10029-094 10861-408 10861-410
Forced Air Circulation Oven Units, Memmert
Ovens Standard Ovens
The Generation 2012 range has natural and forced convection models with a temperature range up to +300 °C.
SingleDISPLAY models; ideal for standard applications. ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), fan speed, exhaust air flap position, programme time, time zones and daylight-saving time. One PT100 temperature sensor in a 4-wire circuit and ethernet interface for reading out the protocol log. Double over-temperature protection including visual alarm: Electronic temperature monitoring with freely adjustable monitoring temperature and mechanical temperatureMore Product Information