4-Acetoxy-3-methoxyphenylacetic acid ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

195761GM
100267-546EA 85 USD
100267-546
4-Acetoxy-3-methoxyphenylacetic acid ≥98%, powder
4-Acetoxy-3-methoxyphenylacetic acid

  • White powder
  • Store in a cool, dry place

MW: 224.22 g/mol
Melting Pt: 138…140 °C
MDL Number: MFCD03424354
CAS Number: 5447-38-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f08fa41372e1b95f0be06919e0db36ce6c4e1fca&vid=f08fa41372e1b95f0be06919e0db36ce6c4e1fca&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR