7-Diethylamino-3-phenylcoumarin ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 7-(Diethylamino)-3-phenylcoumarin

19403250MG
100267-296EA 52.31 USD
100267-296
7-Diethylamino-3-phenylcoumarin ≥98%, powder
7-Diethylamino-3-phenylcoumarin

Yellow Powder


Store in a cool, dry place

Formula: C₁₉H₁₉NO₂
MW: 293.37 g/mol
Melting Pt: 152.1…153.4 °C
MDL Number: MFCD00408422
CAS Number: 84865-19-0
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0625f423655afb04866604455705d4dd8a42205c&vid=0625f423655afb04866604455705d4dd8a42205c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR