Grindometers, Industrial Physics
Supplier: GREAT LAKES GAGES, LLC
VF2110 VF2111 VF2112 VF2113 VF2120 VF2121 VF2122 VF2123
10025-134EA 1168 USD
10025-134 10025-136 10025-138 10025-140 10025-246 10025-248 10025-250 10025-252
Grindometers, Industrial Physics
Mills
These grindometers or gauges of fineness are precision instruments to determine particle size and fineness of many materials like paints, lacquers, pigments, filler, chocolate etc
  • Precision instrument
  • Ergonomic shaped scraper for an easy grip.
  • Many models available
Gauge and bevelled scraper are made of hardened medical grade stainless tool steel and have an accuracy of 2μm.
TQC Grindometers have double grooves with graded slopes graduated in three different parameters: µm (microns), NS (Hegman) and PCU (North).
Certificaciones: ASTM D1210, ASTMMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable