VACUUBRAND VACUU·VIEW and VACUU·VIEW Extended Vacuum Gauges, BrandTech
Supplier: Brandtech
20683220 20683210
10024-956EA 2238.25 USD
10024-956 10024-958
VACUUBRAND VACUU·VIEW and VACUU·VIEW Extended Vacuum Gauges, BrandTech
Pumps Vacuum Gauges
Vacuum gauges with corrosion resistant transducers designed especially for the rigors of laboratory use
  • Mercury-free
  • Digital display for accurate readings
  • Capacitive and Pirani transducers for continuous reading from atmosphere to 10-3 mbar/Torr/hPa
  • Corrosion resistant transducers
The VACUU·VIEW gauge features a capacitive transducer made of alumina ceramic, and is suitable for absolute pressure readings in the rough vacuum range (atmosphere to 0.1mbar/hPa or 0.075Torr). Suitable for rotary evaporators, or any application performed by diaphragm, rootsMore Product Information