Copan Plastic square sampling template, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
T2905C
10024-258CS 179.44 USD
10024-258
Copan Plastic square sampling template, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
Plastic square sampling template used for meat carcass testing and/or enumeration procedures.
Plastic square sampling template used for meat carcass testing and/or enumeration procedures. Can be used in conjunction with the Envirotrans™ Environmental Swabing System.