Reflex Wound Closures, Stoelting
Supplier: Stoelting
59038 59037 59036 59035
100231-346EA 490.38 USD
100231-346 100231-350 100231-348 100231-353
Reflex Wound Closures, Stoelting
First Aid Supplies Wound Clips
The 7 mm clips are a perfect size for mouse surgery.
  • Fast, effective suturing for mice and small rodents
  • Autoclavable
Información de suministro: Wound clip kit includes one applier, one remover, and a box of 100 clips.
Envase: Clips are sold in individual packs of 20, unsterilized.