Triglycerides Standard, Pointe Scietnific
Supplier: HORIBA INSTRUMENTS INCORPORATED
T7531-STD
10022-966EA 88.6 USD
10022-966
Triglycerides Standard, Pointe Scietnific
Standards Biological Standards
Triglycerides Standard is Intended for use as a triglycerides standard for standardization or calibration of enzymatic triglycerides procedures.
  • Standardization or calibration of enzymatic triglycerides procedures
  • Should be used to calibrate the determination of unknown specimens
  • Enzymatic triglycerides assay used
Triglycerides Standard is Intended for use as a triglycerides standard for standardization or calibration of enzymatic triglycerides procedures. The standard should be used to calibrate the determination of unknown specimens using an enzymatic triglycerides assay. Controls should be included to validate the calibration of the assay.
Certificaciones: CE Certified
Información para pedidos: Materials Required but not Provided:
Triglycerides Standards are intended to be used with Pointe Scientific Triglycerides Reagents.