2X YT Agar Plates with Ampicillin 100 and 2% Glucose, Teknova
Supplier: TEKNOVA INC
Y4292
100218-130EA 713.7 USD
100218-130 100218-832
2X YT Agar Plates with Ampicillin 100 and 2% Glucose, Teknova
Media for Microbiology Prepared Media for Microbiology
2X YT is a nutitionally rich medium used for cultivating recombinant strains of E. coli.
  • Contains twice as much yeast extract and 60% more tryptone than LB
  • Sterile
Ingredients (per Liter): 1.6% tryptone (16.0 g/l), 1.0% yeast extract (10.0 g/l), 0.5% sodium chloride (NaCl) (5.0 g/l), 2% glucose, ampicillin (100 µg/ml) and 1.5% agar. Adjust to pH 7.0.
Ambient shipping conditions. Refrigerate to store.