Potassium selenite

Supplier: STREM CHEMICALS INC MS
Danger

93-1971-25G 93-1971-100G
100201-954EA 125.97 USD
100201-954 100210-862
Potassium selenite
Potassium selenite
Formula: K₂O₃Se
MW: 205.165 g/mol
Melting Pt: 875 °C (dec.)
MDL Number: MFCD00049663
CAS Number: 10431-47-7
UN: 2630
ADR: 6.1 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR