Lead(II) oxide 99,999 Pb basis, PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Lead monoxide

93-8219-10G 93-8219-50G
100197-966EA 77.85 USD
100197-966 100207-908
Lead(II) oxide 99,999 Pb basis, PURATREM
Lead(II) oxide
Formula: PbO
MW: 223.2 g/mol
Boiling Pt: 1470
Melting Pt: 888
Density: 9.53
MDL Number: MFCD00011164
CAS Number: 1317-36-8
EINECS: 215-267-0
UN: 2291
ADR: 6.1 III
Merck Index: 13,05431

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR