Hexakis[mu-(acetato-O:O')]-triaqua-mu3-oxotriruthenium(III) ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

44-0700-1G 44-0700-5G
100201-648EA 321.29 USD
100201-648 100201-650
Hexakis[mu-(acetato-O:O')]-triaqua-mu3-oxotriruthenium(III) ≥95%
Hexakis[mu-(acetato-O:O')]-triaqua-mu3-oxotriruthenium(III)
MDL Number: MFCD00269840
CAS Number: 38998-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR