Dichloro[(S)-(-)-2,2',6,6'-tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine][(1S,2S)-(-)-1,2-diphenylethylenediamine]ruthenium(II) ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

44-0386-100MG 44-0386-500MG
100198-776EA 125.97 USD
100198-776 100198-778
Dichloro[(S)-(-)-2,2',6,6'-tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine][(1S,2S)-(-)-1,2-diphenylethylenediamine]ruthenium(II) ≥95%
Dichloro[(S)-(-)-2,2',6,6'-tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine][(1S,2S)-(-)-1,2-diphenylethylenediamine]ruthenium(II)

P-PHOS

MDL Number: MFCD06658134
CAS Number: 821793-36-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR