Brady® B30 Series Printer Ribbon R10000 Series
Supplier: Brady Worldwide
140852 140848 140863 140847 140864 140850 140859 140866 140860 140854 140851
10017-940EA 177.61 USD
10017-940 10017-942 10017-944 10017-946 10017-948 10017-950 10017-952 10017-954 10017-956 10017-958 10017-960 10017-962 10017-964 10017-966 10017-968 10017-970 10017-972 10017-974 10017-976 10017-978 10017-980 10017-982 10017-984 10018-012
Brady® B30 Series Printer Ribbon R10000 Series
Printer Ribbons
B30 Series Printer Ribbon R1000, available in Single Color, Two Color, or Four Color options.
  • Tough, smear-proof ribbon produces long-lasting printed text that can withstand years in industrial areas and outdoor conditions
  • Self-contained B30 Series cartridges drop straight into the printer with a "click"
  • R10000 series ribbons are based on a resin formulation that offers a high performance image with strong solvent and scratch resistance
"Drop & Click" print ribbons for quick and easy supply changes. Ribbons are tough and smear-proof, and theyMore Product Information