DuPont™ Tyvek® 400 Frocks with Mandarin Collar
Supplier: DuPont
TY216SWHSM003000 TY216SWH2X003000 TY216SWHMD003000 TY216SWHXL003000 TY216SWHLG003000 TY216SWH3X003000 TY216SWH4X003000 TY216SWH5X003000
13450-522CS 280.94 USD
13450-522 13450-526 13450-520 13450-524 13450-518 13450-528 13450-530 10016-934
DuPont™ Tyvek® 400 Frocks with Mandarin Collar
Disposable Garments
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Mandarin collar
  • Elastic wrists
  • Extends below hips
  • Front snap closure
  • Serged seams
Frocks feature body-length jacket design with front snap closures for easy donning and doffing. Ideal for applications that do not require the full body coverage of a coverall. Frocks also feature a mandarin collar designed to fit tightly around hooded PAPR and elastic openings at wrists for a tighter fit.
Certificaciones: More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Gender
Garment application Laboratory