DuPont™ Tychem® 2000 Coveralls with Standard Hood and Elastic Wrists and Ankles, Taped Seams
Supplier: DuPont
QC127TYLMD000400 QC127TYLLG000400 QC127TYLXL000400 QC127TYL2X000400 QC127TYL3X000400 QC127TYL4X000400 QC127TYL5X000400
10010-432CS 174.92 USD
10010-432 10010-434 10010-436 10010-438 10010-440 10010-442 10016-424
DuPont™ Tychem® 2000 Coveralls with Standard Hood and Elastic Wrists and Ankles, Taped Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Standard fit hood
  • Elastic wrists and ankles
  • Serged seams
  • Available NAFTA sourced
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Taped seams provide strongMore Product Information