DuPont™ Tychem® 2000 Coveralls with Standard Hood and Elastic Wrists and Ankles, Serged Seams
Supplier: DuPont
QC127SYLLG001200 QC127SYL5X001200 QC127SYLXL001200 QC127SYL3X001200 QC127SYL4X001200 QC127SYLMD001200 QC127SYL2X001200 QC127SYL6X001200 QC127SYL7X001200
10010-212CS 182.98 USD
10010-212 13452-664 10010-214 10010-218 10010-220 10010-210 10010-216 10016-408 10016-410 10016-412 10016-414 10016-416 10016-418 10016-420 10016-422
DuPont™ Tychem® 2000 Coveralls with Standard Hood and Elastic Wrists and Ankles, Serged Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Standard fit hood
  • Elastic wrists and ankles
  • Serged seams
  • Available NAFTA sourced
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Multiple interlockingMore Product Information