DuPont™ Tyvek® 400 Hoods
Supplier: DuPont
TY657SWH00010000
10010-124CS 173.95 USD
10010-124
DuPont™ Tyvek® 400 Hoods
Disposable Garments
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Elastic around face opening
  • Shoulder length
  • Serged seams
  • One size fits most
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durabilityMore Product Information