Alere iScreen® Dip Card, Moderately Complex Multi-Panel Drug Test Kits, Alere Toxicology
Supplier: Alere
IS5M IS10 IS3OCTDIP DOA-134 IS4ADIP IS4MDIP DOA-144 IS8DIP DOA-184 IS9DRUGSCREEN DOA-194 I-DOA-194-191 DOA-2104 DOA-1124-081 DOA-1124-011T-O IS2CTDIP DOA-124 DOA-754 DOA-164-551 IS6BDIP DOA-264 IS6MAPDIP DOA-164 IS5MPDIP DOA-354 IS5ADIP DOA-154 DOA-254 IS5MBDIP DOA-654
89171-330PK 286.31 USD
89171-330 89171-332 10000-572 10000-574 10000-576 10000-578 10000-580 10000-582 10000-584 10000-586 10000-588 10000-590 10000-592 10000-594 10000-596 10000-598 10000-600 10000-602 10000-604 10000-640 10000-642 10000-644 10000-726 10000-728 10000-730 10000-732 10000-734 10000-736 10000-738 10000-740 10000-742 10000-744 10000-746 10000-748 10000-916 10000-918 10000-920 10000-922 10000-924 10000-926 10000-928 10000-930 10000-932 10000-934 10000-936 10000-938 10000-940
Alere iScreen® Dip Card, Moderately Complex Multi-Panel Drug Test Kits, Alere Toxicology
Drug Tests
Rapid Drug Screen for the simultaneous, qualitative detection of single or multiple drug metabolites in human urine
Dip Card Format. Designed to screen single or multiple drug metabolites in one simple dip and read procedure with results in 5 minutes.
Room temperature storage, long shelf-life. Collection cup not included.
Rapid screen for a collection of illicit and prescription drugs. Each kit contains materials for 25 drug tests. Rapid Drug Screens can include Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Buprenorphine (BUP), Cocaine (COC), Methamphetamine (mAMP),More Product Information