Alere iScreen® Dip Card, Moderately Complex Single Panel Drug Test Kits, Alere Toxicology
Supplier: Alere
DMA-114 DAM-114 DBA-114 DBU-114 DPC-114 DPP-114 IS1TCA DTC-114 IS1THCDIP DTH-114 IS1METHDIP IS1BENZODIP DBZ-114 IS1 COC DIP DCO-114 DMT-114 IS1OPI300DIP DMO-114 DMD-114 IS1OPI2000 DOP-114 IS1OXYDIP DOX-114
10000-568PK 37.56 USD
10000-568 10000-570 10000-620 10000-622 10000-624 10000-626 10000-628 10000-630 10000-632 10000-634 10000-636 10000-638 10000-724 10000-892 10000-894 10000-896 10000-898 10000-900 10000-902 10000-904 10000-906 10000-908 10000-910 10000-912 10000-914
Alere iScreen® Dip Card, Moderately Complex Single Panel Drug Test Kits, Alere Toxicology
Drug Tests
Rapid Drug Screen for the simultaneous, qualitative detection of single or multiple drug metabolites in human urine
Dip Card Format. Designed to screen single or multiple drug metabolites in one simple dip and read procedure, with results in five minutes.
Room temperature storage, long shelf-life. Collection cup is not included.
Rapid screen for a collection of illicit and prescription drugs. Each kit contains materials for 25 drug tests. Rapid Drug Screens can include Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Buprenorphine (BUP), Cocaine (COC), Methamphetamine (More Product Information