Alere iCassette® Single Panel, Moderately Complex Drug Test Kits, Alere Toxicology
Supplier: Alere
DBU-102 I-DOX-102 DOX-102 DCO-102
10000-548PK 37.08 USD
10000-548 10000-550 10000-552 10000-554 10000-556 10000-558 10000-560 10000-562 10000-610 10000-882 10000-884 10000-886 10000-888 10000-890
Alere iCassette® Single Panel, Moderately Complex Drug Test Kits, Alere Toxicology
Drug Tests
Rapid Drug Screen for the simultaneous, qualitative detection of single or multiple drug metabolites in human urine
Pipette style drug screen uses three drops of urine per well to detect drug metabolites in urine, with results in five minutes.
Room temperature storage, long shelf-life. Pipette Included. Collection cup is not included.
Rapid screen for a collection of illicit and prescription drugs. Each kit contains materials for 25 or 40 (noted in description) drug tests. Rapid Drug Screens can include Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Buprenorphine (BUP)More Product Information