Alere EZ Split Key® Cup and Key Cup A.D., Moderately Complex Drug Test Kits, Alere Toxicology
Supplier: Alere
DOA-1187-019 DOA-21107-019 DUA-1107-021-19 DOA-2167-019 DOA-167-321-019 DOA-177-161
10000-538PK 252.8 USD
10000-538 10000-540 10000-544 10000-546 10000-714 10000-716 10000-718
Alere EZ Split Key® Cup and Key Cup A.D., Moderately Complex Drug Test Kits, Alere Toxicology
Drug Tests
Rapid Drug Screen for the simultaneous, qualitative detection of single or multiple drug metabolites in human urine
Stand up cup that detects drug metabolites in urine, with results in five minutes. Split specimen cup with key activation. Cup and temperature strip included. SVT option available.
Room temperature storage, long shelf-life.
Rapid screen for a collection of illicit and prescription drugs. Each kit contains materials for 25 drug tests. Rapid Drug Screens can include Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Buprenorphine (BUP),Cocaine (COC),More Product Information