3-Fluoro-4-nitrobenzyl bromide 97%

Supplier: Thermo Scientific Chemicals
Danger

H26193-03 H26193-06
AAH26193-03EA 183.99 USD
AAH26193-03 AAH26193-06
3-Fluoro-4-nitrobenzyl bromide 97%
3-Fluoro-4-nitrobenzyl bromide

Purity: 97% CAS No.: 131858-37-2 Molecular Formula: C7H5BrFNO2 Formula Weight: 234.02 Melting Point: 46-48° UN No.: 3261 MDL No.: MFCD03094237

Formula: C₇H₅BrFNO₂
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD03094237
CAS Number: 131858-37-2
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR