2-(4-Thiazolyl)benzimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T0830-25G T0830-250G
TCT0830-025GEA 36.33 USD
TCT0830-025G TCT0830-250G
2-(4-Thiazolyl)benzimidazole ≥98.0% (by GC, titration analysis)
2-(4-Thiazolyl)benzimidazole

CAS Number: 148-79-8
MDL Number: MFCD00005587
Molecular Formula: C10H7N3S
Molecular Weight: 201.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 305


Product Categories: Synthetic Organic Chemistry, Ligands, Bioscience, Fungicide Ingredients, Anthelmintic Ingredients
Other Product Names / Synonyms: Thiabendazole

Formula: C₁₀H₇N₃S
Melting Pt: 305 °C
MDL Number: MFCD00005587
CAS Number: 148-79-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0af34fbb15db2c1ee46b82245da973528554b5a9&vid=0af34fbb15db2c1ee46b82245da973528554b5a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR