Phenyl propionate ≥97.0%

Supplier: TCI America

Synonyms: Propionic Acid Phenyl Ester

P0509-25G P0509-100G
TCP0509-025GEA 123 USD
TCP0509-025G TCP0509-100G
Phenyl propionate ≥97.0%
Phenyl propionate

CAS Number: 637-27-4
MDL Number: MFCD00053765
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 211
Flash Point (°C): 86
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: Propionic Acid Phenyl Ester

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 211 °C
Density: 1.05 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00053765
CAS Number: 637-27-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d797b4208c744ee8943fc4e2f19786e414da3edb&vid=d797b4208c744ee8943fc4e2f19786e414da3edb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR